missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethylbenzene, 97+%

Catalog No. p-7058860
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43369S0 2 mL
AA43369AB 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43369S0 Supplier Thermo Scientific Chemicals Supplier No. 043369S0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 141-93-5 | C10H14 | 134.222 g/mol

1,3-Diethylbenzene is used as an intermediate and solvent. It is also used in the preparation of divinyl benzene. Further, it is used in powderless etching.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141-93-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00009260
InChI Key AFZZYIJIWUTJFO-UHFFFAOYSA-N
Synonym m-diethylbenzene, benzene, 1,3-diethyl, benzene, m-diethyl, m-ethylethylbenzene, unii-zm2x7i1g8y, zm2x7i1g8y, ethylated benzene, benzene, ethylated, meta-diethylbenzene, polyethylated benzene
PubChem CID 8864
IUPAC Name 1,3-diethylbenzene
SMILES CCC1=CC(=CC=C1)CC

Specifications

Color Colorless
Density 0.86 g/mL
Boiling Point 182°C
Flash Point 55°C (131°F)
Odor Aromatic
Assay Percent Range >97%
Refractive Index 1.496
Quantity 2 mL
UN Number UN2049
Solubility Information Miscible with ethanol,benzene,carbon tetrachloride,ethyl ether and acetone. Immiscible with water and diethylbenzene.
Formula Weight 134.22
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 1,3-Diethylbenzene
Show More Show Less
Hazard Category H226-H304-H315-H319-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYLBENZENE
EINECSNumber 205-511-4
RTECSNumber CZ5620000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only