missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicyclohexylurea 98.0+%, TCI America™

Catalog No. p-7126215
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D044125G 25 g
D0441500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D044125G Supplier TCI America Supplier No. D044125G
Only null left

Chemical Identifiers

CAS 2387-23-7
Molecular Formula C13H24N2O
Molecular Weight (g/mol) 224.348
MDL Number MFCD00003829
InChI Key ADFXKUOMJKEIND-UHFFFAOYSA-N
Synonym n,n'-dicyclohexylurea, dicyclohexylurea, urea, n,n'-dicyclohexyl, unii-zv7823vvim, chembl1458, zv7823vvim, urea,3-dicyclohexyl, urea,n'-dicyclohexyl, n,n inverted exclamation marka-dicyclohexylurea, urea, 1,3-dicyclohexyl
PubChem CID 4277
IUPAC Name 1,3-dicyclohexylurea
SMILES C1CCC(CC1)NC(=O)NC2CCCCC2

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 25 g
Formula Weight 224.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dicyclohexylurea
EINECSNumber (3)-2496&(3)-2985
TSCA Yes