missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%

Catalog No. AAH3663306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3663306 5 g
AAH3663303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3663306 Supplier Thermo Scientific Chemicals Supplier No. H3663306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 286014-38-8 | C15H25BF4N2 | 320.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 286014-38-8
Molecular Formula C15H25BF4N2
Molecular Weight (g/mol) 320.18
MDL Number MFCD04973310
InChI Key CQHXJIHJFMBBQA-UHFFFAOYSA-N
Synonym 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt
PubChem CID 16218169
IUPAC Name 1,3-dicyclohexyl-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1

Specifications

Melting Point 173°C to 177°C
Color Brown to White
Quantity 5 g
Formula Weight 320.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3-Dicyclohexylimidazolium tetrafluoroborate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber NONE

RUO – Research Use Only