missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%

Catalog No. AAH3663303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3663303 1 g
AAH3663306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3663303 Supplier Thermo Scientific Chemicals Supplier No. H3663303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 286014-38-8 | C15H25BF4N2 | 320.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 286014-38-8
Molecular Formula C15H25BF4N2
Molecular Weight (g/mol) 320.18
MDL Number MFCD04973310
InChI Key CQHXJIHJFMBBQA-UHFFFAOYSA-N
Synonym 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt
PubChem CID 16218169
IUPAC Name 1,3-dicyclohexyl-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1

Specifications

Melting Point 173°C to 177°C
Color Brown to White
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 320.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3-Dicyclohexylimidazolium tetrafluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber NONE
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only