missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicyclohexylbenzimidazolium chloride, 95%, Thermo Scientific™

$192.13 - $367.96

Chemical Identifiers

CAS 1034449-15-4
Molecular Formula C19H27ClN2
Molecular Weight (g/mol) 318.89
InChI Key JACUDTKAZHCIPU-UHFFFAOYSA-M
Synonym 1,3-dicyclohexylbenzimidazolium chloride, 1,3-dicyclohexyl-1??,3-benzodiazol-1-ylium chloride, 1,3-dicyclohexyl-1h-benzimidazol-3-ium chloride, 1,3-dicyclohexyl-1h-benzo d imidazol-3-ium chloride, 1,3-dicyclohexyl-2h-1,3-benzodiazol-2-ylium chloride
PubChem CID 16218267
IUPAC Name 1,3-dicyclohexylbenzimidazol-3-ium;chloride
SMILES C1CCC(CC1)N2C=[N+](C3=CC=CC=C32)C4CCCCC4.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC435095000
View Documents Promotion Details
N/A 500mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC435090010
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1034449-15-4
318.89
1,3-dicyclohexylbenzimidazolium chloride, 1,3-dicyclohexyl-1??,3-benzodiazol-1-ylium chloride, 1,3-dicyclohexyl-1h-benzimidazol-3-ium chloride, 1,3-dicyclohexyl-1h-benzo d imidazol-3-ium chloride, 1,3-dicyclohexyl-2h-1,3-benzodiazol-2-ylium chloride
1,3-dicyclohexylbenzimidazol-3-ium;chloride
C19H27ClN2
JACUDTKAZHCIPU-UHFFFAOYSA-M
16218267
C1CCC(CC1)N2C=[N+](C3=CC=CC=C32)C4CCCCC4.[Cl-]

Specifications

1034449-15-4
100.0
Authentic
Glass Bottle
1,3-dicyclohexylbenzimidazolium chloride, 1,3-dicyclohexyl-1??,3-benzodiazol-1-ylium chloride, 1,3-dicyclohexyl-1h-benzimidazol-3-ium chloride, 1,3-dicyclohexyl-1h-benzo d imidazol-3-ium chloride, 1,3-dicyclohexyl-2h-1,3-benzodiazol-2-ylium chloride
C1CCC(CC1)N2C=[N+](C3=CC=CC=C32)C4CCCCC4.[Cl-]
318.89
318.89
1, 3-Dicyclohexylbenzimidazolium chloride
94.0
67.30 to 75.90% C, 8.25 to 9.31% N
95%
C19H27ClN2
JACUDTKAZHCIPU-UHFFFAOYSA-M
1,3-dicyclohexylbenzimidazol-3-ium;chloride
16218267
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only