missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloropropene (cis- and trans- mixture) 92.0+%, TCI America™

Catalog No. p-7126190
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D040525G 25 g
D0405500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D040525G Supplier TCI America Supplier No. D040525G
Only null left

Chemical Identifiers

CAS 542-75-6
Molecular Formula C3H4Cl2
Molecular Weight (g/mol) 110.97
MDL Number MFCD00000985
InChI Key UOORRWUZONOOLO-UPHRSURJSA-N
Synonym 1,3-dichloropropene, trans-1,3-dichloropropene, telone ii, telone, 1,3-dichloropropylene, nemex, trans-1,3-dichloropropylene, dorlone ii, telone c, di-trapex cp
PubChem CID 24726
ChEBI CHEBI:18624
IUPAC Name (1Z)-1,3-dichloroprop-1-ene
SMILES ClC\C=C/Cl

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 g
UN Number 2047
Formula Weight 110.97
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dichloropropene (cis- and trans- mixture)
EINECSNumber (2)-0125
RTECSNumber UC8310000
TSCA Yes