missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloropropane, 95%

Catalog No. AA3668522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3668522 100 g
AA3668514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3668522 Supplier Thermo Scientific Chemicals Supplier No. 03668522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 142-28-9 | C3H6Cl2 | 112.981 g/mol

1,3-Dichloropropane is used as an intermediate in organic synthesis. It is also used as solvent, detergent and chromatographic analysis. Further, it is used to prepare 3,7-dithia-nonanedioic acid by reacting with mercaptoacetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 142-28-9
Molecular Formula C3H6Cl2
Molecular Weight (g/mol) 112.981
MDL Number MFCD00000999
InChI Key YHRUOJUYPBUZOS-UHFFFAOYSA-N
Synonym trimethylene dichloride, propane, 1,3-dichloro, trimethylene chloride, 1,3-dichloro-propane, unii-aj1hq2gucp, ccris 9220, ch2clch2ch2cl, aj1hq2gucp, cl ch2 3cl, 1,3-dichlorpropan
PubChem CID 8881
IUPAC Name 1,3-dichloropropane
SMILES C(CCl)CCl

Specifications

Density 1.190 g/mL
Boiling Point 120°C
Flash Point 20°C (68°F)
Odor Sweet
Refractive Index 1.4481
Quantity 100 g
UN Number UN1993
Solubility Information Miscible with water.
Formula Weight 112.99
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1,3-Dichloropropane
Show More Show Less
Hazard Category H225-H302+H332-H315-H319
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 205-531-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only