missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichlorobenzene 98.0+%, TCI America™

Catalog No. D0333500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0333500G 500 g
D033325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0333500G Supplier TCI America Supplier No. D0333500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 541-73-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000573
InChI Key ZPQOPVIELGIULI-UHFFFAOYSA-N
Synonym m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene
PubChem CID 10943
ChEBI CHEBI:36693
IUPAC Name 1,3-dichlorobenzene
SMILES C1=CC(=CC(=C1)Cl)Cl

Specifications

Melting Point -25°C
Color Yellow
Boiling Point 172°C
Quantity 500 g
UN Number 3082
Formula Weight 147.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dichlorobenzene
EINECSNumber (3)-0041
RTECSNumber CZ4499000
TSCA Yes