missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichlorobenzene, 98%

Catalog No. aaa136880b
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1000 g
250 g
500 g
Catalog No. Quantity
AAA136880B 1000 g
AAA1368830 250 g
AAA1368836 500 g
3 options

Catalog No. AAA136880B

Supplier: Thermo Scientific Chemicals A136880B

Only null left

CAS: 541-73-1 | C6H4Cl2 | 146.998 g/mol

1,3-Dichlorobenzene is used in medicine, and dyes. And it is also used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules1. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichlorobenzene is used in medicine, and dyes. And it is also used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules1. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 541-73-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000573
InChI Key ZPQOPVIELGIULI-UHFFFAOYSA-N
Synonym m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene
PubChem CID 10943
ChEBI CHEBI:36693
IUPAC Name 1,3-dichlorobenzene
SMILES C1=CC(=CC(=C1)Cl)Cl

Specifications

Melting Point -25°C
Density 1.288
Boiling Point 173°C
Flash Point 63°C (145°F)
Refractive Index 1.546
Quantity 1000 g
UN Number UN3082
Beilstein 956618
Merck Index 14,3055
Solubility Information Difficult to mix.
Formula Weight 147
Percent Purity 98%
Chemical Name or Material 1,3-Dichlorobenzene
Show More Show Less
Hazard Category H227-H302-H332
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P374-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 208-792-1
RTECSNumber CZ4499000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only