missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloroacetone, 96%

Catalog No. AAA1414914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1414914 25 g
AAA1414922 100 g
AAA1414936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1414914 Supplier Thermo Scientific Chemicals Supplier No. A1414914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 534-07-6 | C3H4Cl2O | 126.96 g/mol

1,3-Dichloroacetone is used in the synthesis of citric acid. It is used as a solvent and as an intermediate in organic synthesis. Further, it serves as an acceptor substrate in the cross-aldol reaction with donor substrates such as acetone, cyclopentanone and cyclohexanone. It is also used in the preparation of an active pharmaceutical ingredient, famotidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichloroacetone is used in the synthesis of citric acid. It is used as a solvent and as an intermediate in organic synthesis. Further, it serves as an acceptor substrate in the cross-aldol reaction with donor substrates such as acetone, cyclopentanone and cyclohexanone. It is also used in the preparation of an active pharmaceutical ingredient, famotidine.

Solubility
Soluble in water, ethanol and diethyl ether.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents, strong bases and reducing agents.

Chemical Identifiers

CAS 534-07-6
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.96
MDL Number MFCD00000937
InChI Key SUNMBRGCANLOEG-UHFFFAOYSA-N
Synonym 1,3-dichloroacetone, 1,3-dichloro-2-propanone, 1,3-dichloropropanone, s-dichloroacetone, sym-dichloroacetone, bis chloromethyl ketone, 2-propanone, 1,3-dichloro, acetone, 1,3-dichloro, chloromethyl ketone
PubChem CID 10793
IUPAC Name 1,3-dichloropropan-2-one
SMILES ClCC(=O)CCl

Specifications

Melting Point 40°C to 44°C
Density 1.383
pH 4.2
Boiling Point 172°C to 173°C
Flash Point 95°C (203°F)
Odor Pungent
Refractive Index 1.44
Quantity 25 g
UN Number UN2649
Beilstein 605456
Merck Index 14,3052
Solubility Information Soluble in water,ethanol and diethyl ether.
Formula Weight 126.97
Percent Purity 96%
Chemical Name or Material 1,3-Dichloroacetone
Show More Show Less
Hazard Category H300+H310+H330-H314-H335-H340
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318-H341
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P260-P262-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,3-DICHLOROACETONE
EINECSNumber 208-585-6
RTECSNumber UC1430000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only