missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloroacetone, 96%

Catalog No. p-103920
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173630250 25 g Glass bottle
AC173630500 50 g Glass bottle
AC173632500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC173630250 Supplier Thermo Scientific Chemicals Supplier No. 173630250
Only null left

CAS: 534-07-6 | C3H4Cl2O | 126.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 534-07-6
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.96
MDL Number MFCD00000937
InChI Key SUNMBRGCANLOEG-UHFFFAOYSA-N
Synonym 1,3-dichloroacetone, 1,3-dichloro-2-propanone, 1,3-dichloropropanone, s-dichloroacetone, sym-dichloroacetone, bis chloromethyl ketone, 2-propanone, 1,3-dichloro, acetone, 1,3-dichloro, chloromethyl ketone
PubChem CID 10793
IUPAC Name 1,3-dichloropropan-2-one
SMILES ClCC(=O)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 38°C to 42°C
Color White or Colorless
Density 1.3830g/mL
Boiling Point 172°C to 173°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClCH2COCH2Cl
Quantity 25 g
Beilstein 01, 655
Fieser 02,109
Specific Gravity 1.383
Merck Index 15, 3061
Solubility Information Solubility in water: 27.9g/L water (20° c). Other solubilities: 464g/L toluene (20°C),soluble in alcohol,ether and hexane,slightly soluble in acetone,benzene,chloroform,and methanol
Formula Weight 126.97
Percent Purity 96%
Physical Form Crystals
Chemical Name or Material 1, 3-Dichloroacetone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Fatal if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-585-6
RTECSNumber UC1430000
TSCA TSCA

RUO – Research Use Only