missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dichloro-5,5-dimethylhydantoin 97.0+%, TCI America™
Supplier: TCI America D1783100G
Specifications
| 1,3-Dichloro-5,5-dimethylhydantoin | |
| 132°C | |
| C5H6Cl2N2O2 | |
| 100 g | |
| 1,3-dichloro-5,5-dimethylhydantoin, dantoin, dactin, daktin, dichlorantin, omchlor, halane, hydan, hydantoin, dichlorodimethyl, hydan antiseptic | |
| CC1(C(=O)N(C(=O)N1Cl)Cl)C | |
| 197.015 | |
| 197.02 | |
| Crystalline Powder |
| 118-52-5 | |
| White | |
| MFCD00003190 | |
| 3085 | |
| KEQGZUUPPQEDPF-UHFFFAOYSA-N | |
| 1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione | |
| 8360 | |
| ≥97.0% (T) |
Chemical Identifiers
| 118-52-5 | |
| 197.015 | |
| KEQGZUUPPQEDPF-UHFFFAOYSA-N | |
| 8360 | |
| CC1(C(=O)N(C(=O)N1Cl)Cl)C |
| C5H6Cl2N2O2 | |
| MFCD00003190 | |
| 1,3-dichloro-5,5-dimethylhydantoin, dantoin, dactin, daktin, dichlorantin, omchlor, halane, hydan, hydantoin, dichlorodimethyl, hydan antiseptic | |
| 1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione |
Safety and Handling
EINECSNumber : (9)-0580
RTECSNumber : MU0700000
TSCA : Yes