missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-5,5-dimethylhydantoin 97.0+%, TCI America™

Catalog No. D178325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D178325G 25 g
D1783100G 100 g
D1783500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D178325G Supplier TCI America Supplier No. D178325G

Chemical Identifiers

CAS 118-52-5
Molecular Formula C5H6Cl2N2O2
Molecular Weight (g/mol) 197.015
MDL Number MFCD00003190
InChI Key KEQGZUUPPQEDPF-UHFFFAOYSA-N
Synonym 1,3-dichloro-5,5-dimethylhydantoin, dantoin, dactin, daktin, dichlorantin, omchlor, halane, hydan, hydantoin, dichlorodimethyl, hydan antiseptic
PubChem CID 8360
IUPAC Name 1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)N(C(=O)N1Cl)Cl)C

Specifications

Melting Point 132°C
Color White
Quantity 25 g
UN Number 3085
Formula Weight 197.02
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dichloro-5,5-dimethylhydantoin
EINECSNumber (9)-0580
RTECSNumber MU0700000
TSCA Yes