missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-butene (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. D035125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D035125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D035125G Supplier TCI America Supplier No. D035125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 926-57-8
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.99
MDL Number MFCD00000861
InChI Key WLIADPFXSACYLS-DUXPYHPUSA-N
Synonym 1,3-dichlorobutylene, unii-pwg1e5mrlj, 1,3-dichloro-2-butene, trans-1,3-dichlorobutene-2, 1,3-dichlorobut-2-ene, pwg1e5mrlj, 2-butene, 1,3-dichloro-, e, 2-butene, 1,3-dichloro, e-1,3-dichloro-2-butene, 1,3-dichlorobutene-2
PubChem CID 643911
IUPAC Name (2E)-1,3-dichlorobut-2-ene
SMILES C\C(Cl)=C/CCl

Specifications

Color Yellow
Boiling Point 123°C
Quantity 25 g
UN Number 1992
Formula Weight 124.99
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dichloro-2-butene (cis- and trans- mixture)
EINECSNumber (2)-0118
RTECSNumber EM4760000
TSCA Yes