missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-butene, cis + trans, 98%, stab. with BHT

Catalog No. AAL2033406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2033406 5 g
AAL2033414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2033406 Supplier Thermo Scientific Chemicals Supplier No. L2033406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 926-57-8 | C4H6Cl2 | 124.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 926-57-8
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.99
MDL Number MFCD00000861
InChI Key WLIADPFXSACYLS-DUXPYHPUSA-N
Synonym 1,3-dichlorobutylene, unii-pwg1e5mrlj, 1,3-dichloro-2-butene, trans-1,3-dichlorobutene-2, 1,3-dichlorobut-2-ene, pwg1e5mrlj, 2-butene, 1,3-dichloro-, e, 2-butene, 1,3-dichloro, e-1,3-dichloro-2-butene, 1,3-dichlorobutene-2
PubChem CID 643911
SMILES C\C(Cl)=C/CCl

Specifications

Density 1.161
Boiling Point 125°C to 129°C
Flash Point 33°C (91°F)
Refractive Index 1.473
Quantity 5 g
UN Number UN1992
Beilstein 906754
Sensitivity Moisture sensitive
IUPAC Name (2E)-1,3-dichlorobut-2-ene
Formula Weight 125
Percent Purity 98%
Chemical Name or Material 1,3-Dichloro-2-butene, Cis + Trans
Show More Show Less
Hazard Category H226-H301+H331-H312-H314-H335
Hazard Statement GHS H Statement
H331-H226-H302-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 213-138-3
RTECSNumber EM4760000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only