missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 97%

Catalog No. p-7045568
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1117106 5 g
AAL1117114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1117106 Supplier Thermo Scientific Chemicals Supplier No. L1117106
Only null left

CAS: 69304-37-6 | C12H28Cl2OSi2 | 315.425 g/mol

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane is a silylating reagent used for simultaneous protection of 3'- and 5'-hydroxy functions of ribonucleosides. It is used in the analysis of Watson-Crick and Hoogstein base pairing in nucleotides. It is also employed in the formation of ribavirin chemical delivery systems. Further, it serves as a protecting group reagent for open-chain polyhydroxy compounds. In addition to this, it is involved in the preparation of cyclic bridged peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane is a silylating reagent used for simultaneous protection of 3′- and 5′-hydroxy functions of ribonucleosides. It is used in the analysis of Watson-Crick and Hoogstein base pairing in nucleotides. It is also employed in the formation of ribavirin chemical delivery systems. Further, it serves as a protecting group reagent for open-chain polyhydroxy compounds. In addition to this, it is involved in the preparation of cyclic bridged peptides.

Solubility
Miscible with dimethylformamide, hexane, ethyl acetate, chloroform and dichloromethane.

Notes
Incompatible with strong oxidizing agents and water.

Chemical Identifiers

CAS 69304-37-6
Molecular Formula C12H28Cl2OSi2
Molecular Weight (g/mol) 315.425
MDL Number MFCD00009655
InChI Key DDYAZDRFUVZBMM-UHFFFAOYSA-N
Synonym 1,3-dichloro-1,1,3,3-tetraisopropyldisiloxane, 1,3-dichlorotetraisopropyldisiloxane, 1,1,3,3-tetraisopropyl-1,3-dichlorodisiloxane, 1,3-dichloro-1,1,3,3-tetraisopropyl-disiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetrakis 1-methylethyl, 1,1,3,3-tetraisopropyl-1,3-dichlorosiloxane, chloro chlorodiisopropylsilyl oxy diisopropylsilane, chloro-chloro-di propan-2-yl silyl oxy-di propan-2-yl silane, tipdsicl2, tipdsi-cl2
PubChem CID 172404
IUPAC Name chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane
SMILES CC(C)[Si](C(C)C)(O[Si](C(C)C)(C(C)C)Cl)Cl

Specifications

Boiling Point 69°C to 70°C (0.5mmHg)
Quantity 5 g
UN Number UN1760
Beilstein 1934216
Flash Point >110°C (230°F)
Solubility Information Miscible with dimethylformamide,hexane,ethyl acetate,chloroform and dichloromethane.
Formula Weight 315.43
Percent Purity 97%
Refractive Index 1.454
Sensitivity Moisture sensitive
Density 0.986
Chemical Name or Material 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P374-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only