missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 96%

Catalog No. p-105837
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mL
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC215280050 5 mL Glass bottle
1 options

Catalog No. AC215280050

Supplier: Thermo Scientific Chemicals 215280050

Only null left

CAS: 69304-37-6 | C12H28Cl2OSi2 | 315.425 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69304-37-6
MDL Number MFCD00009655
InChI Key DDYAZDRFUVZBMM-UHFFFAOYSA-N
Synonym 1,3-dichloro-1,1,3,3-tetraisopropyldisiloxane, 1,3-dichlorotetraisopropyldisiloxane, 1,1,3,3-tetraisopropyl-1,3-dichlorodisiloxane, 1,3-dichloro-1,1,3,3-tetraisopropyl-disiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetrakis 1-methylethyl, 1,1,3,3-tetraisopropyl-1,3-dichlorosiloxane, chloro chlorodiisopropylsilyl oxy diisopropylsilane, chloro-chloro-di propan-2-yl silyl oxy-di propan-2-yl silane, tipdsicl2, tipdsi-cl2
PubChem CID 172404
IUPAC Name chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane
SMILES CC(C)[Si](C(C)C)(O[Si](C(C)C)(C(C)C)Cl)Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Density 1.0000g/mL
Boiling Point 120.0°C (15.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C12H28Cl2OSi2
Refractive Index 1.4533 to 1.4553
Linear Formula [(CH3)2CH]2Si(Cl)OSi(Cl)[CH(CH3)2]2
Quantity 5 mL
Fieser 16,125; 17,121
Molecular Weight (g/mol) 315.425
Formula Weight 315.42
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 96%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only