Learn More
1,3-Dibromo-5,5-dimethylhydantoin, 98%
CAS: 77-48-5 | C5H6Br2N2O2 | 285.91 g/mol
Supplier: Thermo Scientific Chemicals 169540025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
| 1, 3-Dibromo-5, 5-dimethylhydantoin | |
| 77-48-5 | |
| 100.0 | |
| Cream or Yellow | |
| 0.5% max. | |
| 97.5% min. (Iodometry) | |
| C5H6Br2N2O2 | |
| 2.5 kg | |
| 15,3022 | |
| Solubility in water: 0.1% (25°C). Other solubilities: soluble in chloroform and glacial acetic acid,0.8% in benzene- 0.02% in ccl4 (25°C),insoluble in hexane,decomposes in ethylalcohol and acetone | |
| CC1(C(=O)N(C(=O)N1Br)Br)C | |
| 285.91 | |
| 285.91 | |
| Powder |
| 98% | |
| 97.5 | |
| 187.0°C to 191.0°C | |
| 155°C | |
| Authentic | |
| Plastic bottle | |
| MFCD00003189 | |
| 01,208; 02,108; 12,158; 15,112 | |
| 1,3-dibromo-5,5-dimethylhydantoin, dibromantin, dibromantine, take charge orange, dbdmh, n,n'-dibromodimethylhydantoin, 5,5-dimethyl-1,3-dibromohydantoin, 2,4-imidazolidinedione, 1,3-dibromo-5,5-dimethyl, 1,3-dibromo-5,5-dimethyl-2,4-imidazolidinedione, unii-v9r5f9i7mz | |
| VRLDVERQJMEPIF-UHFFFAOYSA-N | |
| 1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione | |
| 6479 | |
| 98% |
Chemical Identifiers
| 77-48-5 | |
| 285.91 | |
| VRLDVERQJMEPIF-UHFFFAOYSA-N | |
| 6479 | |
| CC1(C(=O)N(C(=O)N1Br)Br)C |
| C5H6Br2N2O2 | |
| MFCD00003189 | |
| 1,3-dibromo-5,5-dimethylhydantoin, dibromantin, dibromantine, take charge orange, dbdmh, n,n'-dibromodimethylhydantoin, 5,5-dimethyl-1,3-dibromohydantoin, 2,4-imidazolidinedione, 1,3-dibromo-5,5-dimethyl, 1,3-dibromo-5,5-dimethyl-2,4-imidazolidinedione, unii-v9r5f9i7mz | |
| 1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 201-030-9
RTECSNumber : MU0686000
TSCA : TSCA
RUO – Research Use Only