missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5,5-dimethylhydantoin, 98%

Catalog No. AC169545000
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169545000 500 g Plastic bottle
AC169540250 25 g Plastic bottle
AC169540025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169545000 Supplier Thermo Scientific Chemicals Supplier No. 169545000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77-48-5 | C5H6Br2N2O2 | 285.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 77-48-5
Molecular Formula C5H6Br2N2O2
Molecular Weight (g/mol) 285.91
MDL Number MFCD00003189
InChI Key VRLDVERQJMEPIF-UHFFFAOYSA-N
Synonym 1,3-dibromo-5,5-dimethylhydantoin, dibromantin, dibromantine, take charge orange, dbdmh, n,n'-dibromodimethylhydantoin, 5,5-dimethyl-1,3-dibromohydantoin, 2,4-imidazolidinedione, 1,3-dibromo-5,5-dimethyl, 1,3-dibromo-5,5-dimethyl-2,4-imidazolidinedione, unii-v9r5f9i7mz
PubChem CID 6479
IUPAC Name 1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)N(C(=O)N1Br)Br)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 187.0°C to 191.0°C
Color Cream or Yellow
Flash Point 155°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Plastic bottle
Quantity 500 g
Fieser 01,208; 02,108; 12,158; 15,112
Merck Index 15,3022
Solubility Information Solubility in water: 0.1% (25°C). Other solubilities: soluble in chloroform and glacial acetic acid,0.8% in benzene- 0.02% in ccl4 (25°C),insoluble in hexane,decomposes in ethylalcohol and acetone
Formula Weight 285.91
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 3-Dibromo-5, 5-dimethylhydantoin, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-030-9
RTECSNumber MU0686000
TSCA TSCA

RUO – Research Use Only