missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminopropane 98.0+%, TCI America™

Catalog No. D011425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D011425ML 25 mL
D0114500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D011425ML Supplier TCI America Supplier No. D011425ML

Chemical Identifiers

CAS 109-76-2
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.127
MDL Number MFCD00008228
InChI Key XFNJVJPLKCPIBV-UHFFFAOYSA-N
Synonym 1,3-diaminopropane, 1,3-propanediamine, trimethylenediamine, 1,3-propylenediamine, propandiamine, alpha,omega-propanediamine, unii-cb3isl56kg, ccris 4054, 1,3-propane diamine, cb3isl56kg
PubChem CID 428
ChEBI CHEBI:15725
IUPAC Name propane-1,3-diamine
SMILES C(CN)CN

Specifications

Color Yellow
Boiling Point 133°C
Quantity 25 mL
UN Number 2734
Formula Weight 74.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diaminopropane
EINECSNumber (2)-0149
RTECSNumber TX6825000
TSCA Yes