missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diaminopropane Dihydrochloride 97.0+%, TCI America™
$89.48 - $277.75
Chemical Identifiers
| CAS | 10517-44-9 |
|---|---|
| Molecular Formula | C3H12Cl2N2 |
| Molecular Weight (g/mol) | 147.04 |
| MDL Number | MFCD00012525 |
| InChI Key | HYOCSVGEQMCOGE-UHFFFAOYSA-N |
| Synonym | Trimethylenediamine Dihydrochloride, 1,3-Propanediamine Dihydrochloride |
| PubChem CID | 165887 |
| IUPAC Name | dihydrogen propane-1,3-diamine dichloride |
| SMILES | [H+].[H+].[Cl-].[Cl-].NCCCN |
Chemical Identifiers
| 10517-44-9 | |
| 147.04 | |
| HYOCSVGEQMCOGE-UHFFFAOYSA-N | |
| 165887 | |
| [H+].[H+].[Cl-].[Cl-].NCCCN |
| C3H12Cl2N2 | |
| MFCD00012525 | |
| Trimethylenediamine Dihydrochloride, 1,3-Propanediamine Dihydrochloride | |
| dihydrogen propane-1,3-diamine dichloride |
Specifications
| 10517-44-9 | |
| White | |
| MFCD00012525 | |
| Trimethylenediamine Dihydrochloride, 1,3-Propanediamine Dihydrochloride | |
| [H+].[H+].[Cl-].[Cl-].NCCCN | |
| 147.04 | |
| 147.04 | |
| Crystalline Powder |
| 253°C | |
| C3H12Cl2N2 | |
| 25 g | |
| HYOCSVGEQMCOGE-UHFFFAOYSA-N | |
| dihydrogen propane-1,3-diamine dichloride | |
| 165887 | |
| ≥97.0% (T) | |
| 1,3-Diaminopropane Dihydrochloride |
Safety and Handling
EINECSNumber : (2)-0149
RTECSNumber : TX7392500
TSCA : No