missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminopropane Dihydrochloride (Low water content) 98.0+%, TCI America™

Catalog No. D52531G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52531G 1 g
D52535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52531G Supplier TCI America Supplier No. D52531G
Only null left

Chemical Identifiers

CAS 10517-44-9
Molecular Formula C3H12Cl2N2
Molecular Weight (g/mol) 147.04
MDL Number MFCD00012525
InChI Key HYOCSVGEQMCOGE-UHFFFAOYSA-N
Synonym Trimethylenediamine Dihydrochloride, 1,3-Propanediamine Dihydrochloride
PubChem CID 165887
IUPAC Name dihydrogen propane-1,3-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCCN

Specifications

Melting Point 253°C
Color White
Quantity 1 g
Formula Weight 147.04
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diaminopropane Dihydrochloride (Low water content)
EINECSNumber (2)-0149
RTECSNumber TX7392500
TSCA No