missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminopropane, 99%

Catalog No. AC112350500
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112350500 50 mL Glass bottle
AC112352500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112350500 Supplier Thermo Scientific Chemicals Supplier No. 112350500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 109-76-2 | C3H10N2 | 74.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-76-2
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.13
MDL Number MFCD00008228
InChI Key XFNJVJPLKCPIBV-UHFFFAOYSA-N
Synonym 1,3-Propanediamine, Trimethylenediamine
PubChem CID 5942
ChEBI CHEBI:16841
IUPAC Name propane-1,3-diamine
SMILES NCCCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -12°C
Density 0.8800g/mL
Boiling Point 140°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4565 to 1.4585
Linear Formula H2N(CH2)3NH2
Quantity 50 mL
Beilstein 04, 261
Fieser 13,160
Specific Gravity 0.88
Solubility Information Solubility in water: soluble. Other solubilities: miscible with almost all common organic solvents
Formula Weight 74.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 3-Diaminopropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Fatal in contact with skin.
Flammable liquid and vapor.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breat
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rin
EINECSNumber 203-702-7

RUO – Research Use Only