missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminoguanidine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7127009
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D19125G 5 g
D191225G 25 g
2 options

Catalog No. D19125G

Supplier: TCI America D19125G

Only null left

Chemical Identifiers

CAS 36062-19-8
Molecular Formula CH8ClN5
Molecular Weight (g/mol) 125.56
MDL Number MFCD00012948
InChI Key HAZRIBSLCUYMQP-UHFFFAOYSA-N
Synonym 1,3-diaminoguanidine hydrochloride, unii-014y506oeg, 1,3-diaminoguanidine monohydrochloride, carbonimidic dihydrazide, hydrochloride, n,n'-diaminoguanidine monohydrochloride, n,n'-diaminoguanidine, carbonimidic dihydrazide, hydrochloride 1:?, 1,2-diaminoguanidine hydrochloride, n,n'-diaminoguandine, hydrochloride, pubchem20888
PubChem CID 9566041
IUPAC Name 1,2-diaminoguanidine;hydrochloride
SMILES C(=NN)(N)NN.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 125.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diaminoguanidine Hydrochloride