missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diamino-2-propanol 97.0+%, TCI America™

Catalog No. p-7130568
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
H04975G 5 g
H049725G 25 g
2 options

Catalog No. H04975G

Supplier: TCI America H04975G

Only null left

Chemical Identifiers

CAS 616-29-5
Molecular Formula C3H10N2O
Molecular Weight (g/mol) 90.126
MDL Number MFCD00008142
InChI Key UYBWIEGTWASWSR-UHFFFAOYSA-N
Synonym 1,3-diamino-2-propanol, 1,3-diamino-2-hydroxypropane, diaminoisopropanol, 2-hydroxy-1,3-diaminopropane, 2-propanol, 1,3-diamino, 2-hydroxy-1,3-propanediamine, 1,3-diamino-2-hydroxy-propane, 2-propanol,3-diamino, acmc-1b1xv, 1,3 diamino-propan-2-ol
PubChem CID 61157
IUPAC Name 1,3-diaminopropan-2-ol
SMILES C(C(CN)O)N

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 114°C
Quantity 5 g
UN Number 3259
Formula Weight 90.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,3-Diamino-2-propanol
EINECSNumber (2)-0315
TSCA Yes