missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diamino-2-propanol, 97%

CAS: 616-29-5 | C3H10N2O | 90.126 g/mol

Supplier:  Thermo Scientific Chemicals A1961809

 View more versions of this product

Catalog No. AAA1961809


Only null left
Explore available promotions

Description

Description

It finds its application as the formaldehyde absorbent, carbon dioxide and acid gas absorption. It is also used as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as the formaldehyde absorbent, carbon dioxide and acid gas absorption. It is also used as a pharmaceutical intermediate and in organic synthesis.

Solubility
Very soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

1,3-Diamino-2-propanol
40°C to 45°C
>110°C (230°F)
MFCD00008142
741859
Very soluble in water.
C(C(CN)O)N
90.126
90.13
616-29-5
235°C
C3H10N2O
10 g
1,3-diamino-2-propanol, 1,3-diamino-2-hydroxypropane, diaminoisopropanol, 2-hydroxy-1,3-diaminopropane, 2-propanol, 1,3-diamino, 2-hydroxy-1,3-propanediamine, 1,3-diamino-2-hydroxy-propane, 2-propanol,3-diamino, acmc-1b1xv, 1,3 diamino-propan-2-ol
UYBWIEGTWASWSR-UHFFFAOYSA-N
1,3-diaminopropan-2-ol
61157
97%

Chemical Identifiers

616-29-5
90.126
UYBWIEGTWASWSR-UHFFFAOYSA-N
61157
C(C(CN)O)N
C3H10N2O
MFCD00008142
1,3-diamino-2-propanol, 1,3-diamino-2-hydroxypropane, diaminoisopropanol, 2-hydroxy-1,3-diaminopropane, 2-propanol, 1,3-diamino, 2-hydroxy-1,3-propanediamine, 1,3-diamino-2-hydroxy-propane, 2-propanol,3-diamino, acmc-1b1xv, 1,3 diamino-propan-2-ol
1,3-diaminopropan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-474-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only