missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diacetylbenzene 98.0+%, TCI America™

Catalog No. p-7127404
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
D26171G 1 g
D26175G 5 g
2 options

Catalog No. D26171G

Supplier: TCI America D26171G

Only null left

Chemical Identifiers

CAS 6781-42-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008740
InChI Key VCHOFVSNWYPAEF-UHFFFAOYSA-N
Synonym 1,3-diacetylbenzene, 1,1'-1,3-phenylene diethanone, ethanone, 1,1'-1,3-phenylene bis, m-diacetylbenzene, benzene-1,3-bis acetyl, m-acetylacetophenone, m-acetyl acetophenone, 1-3-acetylphenyl ethanone, 1-3-acetylphenyl ethan-1-one, m-diacetyl benzene
PubChem CID 23229
IUPAC Name 1-(3-acetylphenyl)ethan-1-one
SMILES CC(=O)C1=CC(=CC=C1)C(C)=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,3-Diacetylbenzene