missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diacetoxyacetone 98.0+%, TCI America™
$80.85 - $390.80
Chemical Identifiers
| CAS | 6946-10-7 |
|---|---|
| Molecular Formula | C7H10O5 |
| Molecular Weight (g/mol) | 174.15 |
| MDL Number | MFCD01310655 |
| InChI Key | PZVCVSQSQHGBNE-UHFFFAOYSA-N |
| Synonym | 1,3-Diacetoxy-2-propanone |
| PubChem CID | 243418 |
| IUPAC Name | (3-acetyloxy-2-oxopropyl) acetate |
| SMILES | CC(=O)OCC(=O)COC(=O)C |
Chemical Identifiers
| 6946-10-7 | |
| 174.15 | |
| PZVCVSQSQHGBNE-UHFFFAOYSA-N | |
| 243418 | |
| CC(=O)OCC(=O)COC(=O)C |
| C7H10O5 | |
| MFCD01310655 | |
| 1,3-Diacetoxy-2-propanone | |
| (3-acetyloxy-2-oxopropyl) acetate |
Specifications
| 6946-10-7 | |
| C7H10O5 | |
| 5 g | |
| PZVCVSQSQHGBNE-UHFFFAOYSA-N | |
| (3-acetyloxy-2-oxopropyl) acetate | |
| 243418 | |
| ≥98.0% (GC) | |
| 1,3-Diacetoxyacetone |
| 50°C | |
| MFCD01310655 | |
| 1,3-Diacetoxy-2-propanone | |
| CC(=O)OCC(=O)COC(=O)C | |
| 174.15 | |
| 174.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No