missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butyl-1,3,2-diazaphospholidine 2-Oxide 95.0+%, TCI America™

Catalog No. D38465G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38465G 5 g
D38461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38465G Supplier TCI America Supplier No. D38465G
Only null left

Chemical Identifiers

CAS 854929-38-7
Molecular Formula C10H22N2OP+
Molecular Weight (g/mol) 217.273
InChI Key RBABEMWOZLGIIE-UHFFFAOYSA-N
PubChem CID 11816821
IUPAC Name 1,3-ditert-butyl-1,3,2-diazaphospholidin-2-ium 2-oxide
SMILES CC(C)(C)N1CCN([P+]1=O)C(C)(C)C

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 5 g
Formula Weight 218.28
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di-tert-butyl-1,3,2-diazaphospholidine 2-Oxide