missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-o-tolylguanidine 99.0+%, TCI America™

Catalog No. p-7126518
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D095325G 25 g
D0953500G 500 g
2 options

Catalog No. D095325G

Supplier: TCI America D095325G

Only null left

Chemical Identifiers

CAS 97-39-2
Molecular Formula C15H17N3
Molecular Weight (g/mol) 239.322
MDL Number MFCD00008513
InChI Key OPNUROKCUBTKLF-UHFFFAOYSA-N
Synonym 1,3-di-o-tolylguanidine, di-o-tolylguanidine, dotg, vulkacite dotg, ditolylguanidine, 1,3-ditolylguanidine, vulkacit dotg, eveite dotg, vulkacit dotg/c, nocceler dt
PubChem CID 7333
IUPAC Name 1,2-bis(2-methylphenyl)guanidine
SMILES CC1=CC=CC=C1NC(=NC2=CC=CC=C2C)N

Specifications

Melting Point 175°C
Color White
Quantity 25 g
Formula Weight 239.32
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di-o-tolylguanidine
EINECSNumber (3)-2190&(9)-1870
RTECSNumber MF1400000
TSCA Yes