missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-BOC-2-(trifluoromethylsulfonyl)guanidine, 96%

Catalog No. AC399750010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399750010 1 g Glass Bottle
AC399750050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399750010 Supplier Thermo Scientific Chemicals Supplier No. 399750010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 207857-15-6 | C12H20F3N3O6S | 391.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207857-15-6
Molecular Formula C12H20F3N3O6S
Molecular Weight (g/mol) 391.36
InChI Key GOQZIPJCBUYLIR-UHFFFAOYSA-N
Synonym 1,3-di-boc-2-trifluoromethylsulfonyl guanidine, goodman's reagent, n,n'-di-boc-n-triflylguanidine, 1,3-bis tert-butoxycarbonyl-2-trifluoromethanesulfonyl guanidine, 1,3-bis tert-butoxycarbonyl-2-trifluoromethylsulfonyl guanidine, 1,3-di-boc-2-trifluoromethanesulfonyl guanidine, tert-butyl n-n-2-methylpropan-2-yl oxycarbonyl-n'-trifluoromethylsulfonyl carbamimidoyl carbamate, tert-butyl n-tert-butoxycarbonyl amino trifluoromethanesulfonylimino methyl carbamate, tert-butyl n-tert-butoxycarbonyl-n'-trifluoromethyl sulphonyl carbamimidoyl carbamate
PubChem CID 3694570
IUPAC Name tert-butyl N-[N-[(2-methylpropan-2-yl)oxycarbonyl]-N'-(trifluoromethylsulfonyl)carbamimidoyl]carbamate
SMILES CC(C)(C)OC(=O)NC(=NS(=O)(=O)C(F)(F)F)NC(=O)OC(C)(C)C

Specifications

Melting Point 111°C to 113°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 391.36
Percent Purity 98%
Chemical Name or Material 1, 3-Di-BOC-2-(trifluoromethylsulfonyl)guanidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only