missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di(4-pyridyl)propane 97.0+%, TCI America™

Catalog No. D0938500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0938500G 500 g
D093825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0938500G Supplier TCI America Supplier No. D0938500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17252-51-6
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD00038046
InChI Key OGNCVVRIKNGJHQ-UHFFFAOYSA-N
Synonym 4,4'-trimethylenedipyridine, 1,3-di 4-pyridyl propane, 1,3-bis 4-pyridyl propane, pyridine, 4,4'-1,3-propanediyl bis, 4,4'-propane-1,3-diyldipyridine, 4-3-pyridin-4-ylpropyl pyridine, 4-3-4-pyridyl propyl pyridine, 4-3-pyridin-4-yl propyl pyridine, pyridine, 4,4'-trimethylenedi, 4,4'-propane-1,3-diyl dipyridine
PubChem CID 87019
IUPAC Name 4-(3-pyridin-4-ylpropyl)pyridine
SMILES C1=CN=CC=C1CCCC2=CC=NC=C2

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 162°C
Quantity 500 g
Formula Weight 198.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di(4-pyridyl)propane