missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-4-piperidylpropane 97.0+%, TCI America™

Catalog No. D0293500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0293500G 500 g
D029325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0293500G Supplier TCI America Supplier No. D0293500G

Chemical Identifiers

CAS 16898-52-5
Molecular Formula C13H26N2
Molecular Weight (g/mol) 210.365
MDL Number MFCD00038013
InChI Key OXEZLYIDQPBCBB-UHFFFAOYSA-N
Synonym 1,3-bis 4-piperidyl propane, 4,4'-trimethylenedipiperidine, 1,3-di-4-piperidylpropane, piperidine, 4,4'-1,3-propanediyl bis, di-pip, 4-3-piperidin-4-yl propyl piperidine, 4-3-4-piperidyl propyl piperidine, unii-eu53ec219i, 1,3-di 4-piperidyl propane, 1,3-bis 4-piperidinyl propane
PubChem CID 85631
IUPAC Name 4-(3-piperidin-4-ylpropyl)piperidine
SMILES C1CNCCC1CCCC2CCNCC2

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 500 g
Formula Weight 210.37
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di-4-piperidylpropane
EINECSNumber (5)-5660
TSCA Yes