missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di(1-adamantyl)imidazolium Tetrafluoroborate 96.0+%, TCI America™

Catalog No. D36211G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36211G Supplier TCI America Supplier No. D36211G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 286014-42-4
Molecular Formula C23H33BF4N2
Molecular Weight (g/mol) 424.335
MDL Number MFCD04973311
InChI Key KVWCCJYLKCSVME-UHFFFAOYSA-N
Synonym 1,3-bis 1-adamantyl imidazolium tetrafluoroborate, 1,3-di 1-adamantyl imidazolium tetrafluoroborate, 1,3-di adamantan-1-yl-1h-imidazol-3-ium tetrafluoroborate, 1,3-bis adamantan-1-yl-1??-imidazol-1-ylium tetrafluoroborate, iad.hbf4, acmc-209h3n, 1,3-diadamantyl-imidazolium tetrafluoroborate, 1,3-di 1-adamantyl imidazoliumtetrafluoroborate, 1,3-bis tricyclo 3.3.1.13,7 dec-1-yl-1h-imidazolium tetrafluoroborate
PubChem CID 16218105
IUPAC Name 1,3-bis(1-adamantyl)imidazol-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1C2CC3CC1CC(C2)(C3)N4C=C[N+](=C4)C56CC7CC(C5)CC(C7)C6

Specifications

Melting Point 282°C
Color Yellow
Quantity 1 g
UN Number 1759
Formula Weight 424.33
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di(1-adamantyl)imidazolium Tetrafluoroborate