missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Cyclopentanedione 98.0+%, TCI America™
$67.11 - $218.10
Chemical Identifiers
| CAS | 3859-41-4 |
|---|---|
| Molecular Formula | C5H6O2 |
| Molecular Weight (g/mol) | 98.10 |
| MDL Number | MFCD00001405 |
| InChI Key | LOGSONSNCYTHPS-UHFFFAOYSA-N |
| Synonym | 1,3-cyclopentanedione, 1,3-cyclopentadione, 1.3-cyclopentanedione, cyclopentan-1,3-dion, 1,3cyclopentadione, 1,3 cyclopentadione, 1,3-cyclopentandione, 1,3-dioxocyclopentane, cyclopentan-1,3-dione, 1,3-cyclopentane dione |
| PubChem CID | 77466 |
| ChEBI | CHEBI:41456 |
| IUPAC Name | cyclopentane-1,3-dione |
| SMILES | O=C1CCC(=O)C1 |
Chemical Identifiers
| 3859-41-4 | |
| 98.10 | |
| LOGSONSNCYTHPS-UHFFFAOYSA-N | |
| 77466 | |
| cyclopentane-1,3-dione |
| C5H6O2 | |
| MFCD00001405 | |
| 1,3-cyclopentanedione, 1,3-cyclopentadione, 1.3-cyclopentanedione, cyclopentan-1,3-dion, 1,3cyclopentadione, 1,3 cyclopentadione, 1,3-cyclopentandione, 1,3-dioxocyclopentane, cyclopentan-1,3-dione, 1,3-cyclopentane dione | |
| CHEBI:41456 | |
| O=C1CCC(=O)C1 |
Specifications
| 3859-41-4 | |
| C5H6O2 | |
| 1 g | |
| LOGSONSNCYTHPS-UHFFFAOYSA-N | |
| cyclopentane-1,3-dione | |
| 77466 | |
| 98.10 | |
| Crystalline Powder |
| 150°C | |
| MFCD00001405 | |
| 1,3-cyclopentanedione, 1,3-cyclopentadione, 1.3-cyclopentanedione, cyclopentan-1,3-dion, 1,3cyclopentadione, 1,3 cyclopentadione, 1,3-cyclopentandione, 1,3-dioxocyclopentane, cyclopentan-1,3-dione, 1,3-cyclopentane dione | |
| O=C1CCC(=O)C1 | |
| 98.10 | |
| CHEBI:41456 | |
| ≥98.0% (GC) | |
| 1,3-Cyclopentanedione |
Safety and Handling
TSCA : No