missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Cyclopentanedione 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 150°C |
| Quantity | 5 g |
| Formula Weight | 98.10 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,3-Cyclopentanedione |
Safety and Handling
| TSCA | No |