missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclopentanedione, 99%

CAS: 3859-41-4 | C5H6O2 | 98.10 g/mol

$285.17 - $942.19

Chemical Identifiers

CAS 3859-41-4
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00001405
InChI Key LOGSONSNCYTHPS-UHFFFAOYSA-N
PubChem CID 77466
ChEBI CHEBI:41456
IUPAC Name cyclopentane-1,3-dione
SMILES O=C1CCC(=O)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC175720050
SDS View Documents
Thermo Scientific Chemicals
175720050
5 g Glass bottle
Each for $285.17
Only null left
 
AC175720250
SDS View Documents
Thermo Scientific Chemicals
175720250
25 g Glass bottle
Each for $942.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3859-41-4
98.10
LOGSONSNCYTHPS-UHFFFAOYSA-N
CHEBI:41456
O=C1CCC(=O)C1
C5H6O2
MFCD00001405
77466
cyclopentane-1,3-dione

Specifications

3859-41-4
100.0
Ochre to Brown
99%
C5H6O2
MFCD00001405
1,3-cyclopentanedione, 1,3-cyclopentadione, 1.3-cyclopentanedione, cyclopentan-1,3-dion, 1,3cyclopentadione, 1,3 cyclopentadione, 1,3-cyclopentandione, 1,3-dioxocyclopentane, cyclopentan-1,3-dione, 1,3-cyclopentane dione
O=C1CCC(=O)C1
98.10
CHEBI:41456
99%
1, 3-Cyclopentanedione
98.5
148°C to 153°C
Authentic
Glass bottle
C5H6(=O)2
5 g
LOGSONSNCYTHPS-UHFFFAOYSA-N
cyclopentane-1,3-dione
77466
98.1
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-372-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only