missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclopentanedione, 98%

CAS: 3859-41-4 | C5H6O2 | 98.10 g/mol

$92.68 - $301.86

Chemical Identifiers

CAS 3859-41-4
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00001405
InChI Key LOGSONSNCYTHPS-UHFFFAOYSA-N
PubChem CID 77466
ChEBI CHEBI:41456
IUPAC Name cyclopentane-1,3-dione
SMILES O=C1CCC(=O)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1033603
View Documents
Thermo Scientific Chemicals
L1033603
1 g
Each for $92.68
Only null left
 
AAL1033606
View Documents
Thermo Scientific Chemicals
L1033606
5 g
Each for $301.86
Only null left
 
Description

Description

1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

3859-41-4
98.10
LOGSONSNCYTHPS-UHFFFAOYSA-N
CHEBI:41456
O=C1CCC(=O)C1
C5H6O2
MFCD00001405
77466
cyclopentane-1,3-dione

Specifications

3859-41-4
C5H6O2
1 g
1,3-cyclopentanedione, 1,3-cyclopentadione, 1.3-cyclopentanedione, cyclopentan-1,3-dion, 1,3cyclopentadione, 1,3 cyclopentadione, 1,3-cyclopentandione, 1,3-dioxocyclopentane, cyclopentan-1,3-dione, 1,3-cyclopentane dione
LOGSONSNCYTHPS-UHFFFAOYSA-N
cyclopentane-1,3-dione
77466
98.1
1,3-Cyclopentanedione
151°C to 153°C
MFCD00001405
1362728
Very soluble in water.
O=C1CCC(=O)C1
98.10
CHEBI:41456
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-372-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only