missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Cyclopentanedione, 98%
Description
1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.
Solubility
Very soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.
Solubility
Very soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 151°C to 153°C |
| Quantity | 5 g |
| Beilstein | 1362728 |
| Synonym | 1,3-cyclopentanedione, 1,3-cyclopentadione, 1.3-cyclopentanedione, cyclopentan-1,3-dion, 1,3cyclopentadione, 1,3 cyclopentadione, 1,3-cyclopentandione, 1,3-dioxocyclopentane, cyclopentan-1,3-dione, 1,3-cyclopentane dione |
| Solubility Information | Very soluble in water. |
| Formula Weight | 98.1 |
| Percent Purity | 98% |
| Chemical Name or Material | 1,3-Cyclopentanedione |
Safety and Handling
| EINECSNumber | 223-372-8 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only