missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclopentanedione, 98%

Catalog No. AAL1033603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1033603 1 g
AAL1033606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1033603 Supplier Thermo Scientific Chemicals Supplier No. L1033603
Only null left

CAS: 3859-41-4 | C5H6O2 | 98.10 g/mol

1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 3859-41-4
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00001405
InChI Key LOGSONSNCYTHPS-UHFFFAOYSA-N
PubChem CID 77466
ChEBI CHEBI:41456
IUPAC Name cyclopentane-1,3-dione
SMILES O=C1CCC(=O)C1

Specifications

Melting Point 151°C to 153°C
Quantity 1 g
Beilstein 1362728
Synonym 1,3-cyclopentanedione, 1,3-cyclopentadione, 1.3-cyclopentanedione, cyclopentan-1,3-dion, 1,3cyclopentadione, 1,3 cyclopentadione, 1,3-cyclopentandione, 1,3-dioxocyclopentane, cyclopentan-1,3-dione, 1,3-cyclopentane dione
Solubility Information Very soluble in water.
Formula Weight 98.1
Percent Purity 98%
Chemical Name or Material 1,3-Cyclopentanedione
EINECSNumber 223-372-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only