missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanedione, 97%, Thermo Scientific™

Chemical Identifiers

CAS 504-02-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00001585
InChI Key HJSLFCCWAKVHIW-UHFFFAOYSA-N
Synonym 1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
PubChem CID 10434
ChEBI CHEBI:17766
IUPAC Name cyclohexane-1,3-dione
SMILES C1CC(=O)CC(=O)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC111171000
SDS View Documents
Thermo Scientific Chemicals
111171000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC111175000
SDS View Documents
Thermo Scientific Chemicals
111175000
500g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

504-02-9
112.13
HJSLFCCWAKVHIW-UHFFFAOYSA-N
10434
cyclohexane-1,3-dione
C6H8O2
MFCD00001585
1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
CHEBI:17766
C1CC(=O)CC(=O)C1

Specifications

504-02-9
100.0
1% max. Sodium chloride (Argentometry)
C6H8O2
MFCD00001585
07, 554
15, 3196
Solubility in water: soluble. Other solubilities: soluble in alcohol,chloroform,acetone and,boiling benzene,slightly soluble in ether,carbon disulfide and,petroleum ether
C1CC(=O)CC(=O)C1
112.13
CHEBI:17766
97%
96.0
Authentic
Plastic bottle
C6H8(=O)2
100g
01,178
1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
HJSLFCCWAKVHIW-UHFFFAOYSA-N
cyclohexane-1,3-dione
10434
112.13
1, 3-Cyclohexanedione, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-980-0

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only