missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanedione, 97%

Catalog No. AC111171000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC111171000 100 g
AC111175000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111171000 Supplier Thermo Scientific Chemicals Supplier No. 111171000
Only null left

CAS: 504-02-9 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 504-02-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00001585
InChI Key HJSLFCCWAKVHIW-UHFFFAOYSA-N
Synonym 1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
PubChem CID 10434
ChEBI CHEBI:17766
IUPAC Name cyclohexane-1,3-dione
SMILES C1CC(=O)CC(=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point >101°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 1% max. Sodium chloride (Argentometry)
Packaging Plastic bottle
Linear Formula C6H8(=O)2
Quantity 100 g
Beilstein 07, 554
Fieser 01,178
Merck Index 15, 3196
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,chloroform,acetone and,boiling benzene,slightly soluble in ether,carbon disulfide and,petroleum ether
Formula Weight 112.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 3-Cyclohexanedione, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-980-0
TSCA TSCA

RUO – Research Use Only