missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanedione, 97%, may cont. up to 1% NaCl

CAS: 504-02-9 | C6H8O2 | 112.128 g/mol

$50.15 - $598.44

Chemical Identifiers

CAS 504-02-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001585
InChI Key HJSLFCCWAKVHIW-UHFFFAOYSA-N
Synonym 1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
PubChem CID 10434
ChEBI CHEBI:17766
IUPAC Name cyclohexane-1,3-dione
SMILES C1CC(=O)CC(=O)C1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1110914
View Documents
Thermo Scientific Chemicals
A1110914
25 g
Each for $50.15
Only null left
 
AAA1110918
View Documents
Thermo Scientific Chemicals
A1110918
50 g
Each for $56.85
Only null left
 
AAA1110922
View Documents
Thermo Scientific Chemicals
A1110922
100 g
Each for $97.45
Only null left
 
AAA111090B
View Documents
Thermo Scientific Chemicals
A111090B
1000 g
Each for $598.44
Only null left
 
Description

Description

It is used as a nitisinone intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a nitisinone intermediate.

Solubility
Soluble in water (155 g/L) at 20°C, alcohol, chloroform, acetone, boiling benzene, ether (slightly), carbon disulfide (slightly), and petr ether (slightly).

Notes
Air & Light Sensitive. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
Specifications

Chemical Identifiers

504-02-9
112.128
HJSLFCCWAKVHIW-UHFFFAOYSA-N
10434
cyclohexane-1,3-dione
C6H8O2
MFCD00001585
1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
CHEBI:17766
C1CC(=O)CC(=O)C1

Specifications

504-02-9
1.1
MFCD00001585
385888
14,3178
Soluble in water (155g/L) at 20°C,alcohol,chloroform,acetone,boiling benzene,ether (slightly),carbon disulfide (slightly),and petr ether (slightly).
C1CC(=O)CC(=O)C1
112.128
CHEBI:17766
≥97%
1,3-Cyclohexanedione
101°C to 105°C
C6H8O2
25 g
Light sensitive
1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
HJSLFCCWAKVHIW-UHFFFAOYSA-N
cyclohexane-1,3-dione
10434
112.13
May contain up to 1% NaCl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 207-980-0

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only