missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Cyclohexanediol (cis- and trans- mixture) 98.0+%, TCI America™
Supplier: TCI America C048125G
Specifications
| 1,3-Cyclohexanediol (cis- and trans- mixture) | |
| White | |
| C6H12O2 | |
| 25 g | |
| RLMGYIOTPQVQJR-UHFFFAOYSA-N | |
| cyclohexane-1,3-diol | |
| 10433 | |
| ≥98.0% (GC) |
| 504-01-8 | |
| 247°C | |
| MFCD00039458 | |
| 1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7 | |
| C1CC(CC(C1)O)O | |
| 116.16 | |
| 116.16 | |
| Crystalline Powder |
Chemical Identifiers
| 504-01-8 | |
| 116.16 | |
| RLMGYIOTPQVQJR-UHFFFAOYSA-N | |
| 10433 | |
| C1CC(CC(C1)O)O |
| C6H12O2 | |
| MFCD00039458 | |
| 1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7 | |
| cyclohexane-1,3-diol |
Safety and Handling
EINECSNumber : (3)-2310
TSCA : No