missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanediol, cis + trans, 98%

Catalog No. aal0411006
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAL0411006 5 g
AAL0411014 25 g
2 options

Catalog No. AAL0411006

Supplier: Thermo Scientific Chemicals L0411006

Only null left

CAS: 504-01-8 | C6H12O2 | 116.16 g/mol

1,3-Cyclohexanediol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclohexanediol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 504-01-8
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00039458
InChI Key RLMGYIOTPQVQJR-UHFFFAOYSA-N
Synonym 1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7
PubChem CID 10433
IUPAC Name cyclohexane-1,3-diol
SMILES C1CC(CC(C1)O)O

Specifications

Melting Point ∼30°C
Boiling Point 246°C to 247°C
Quantity 5 g
Beilstein 2036654
Solubility Information Soluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 1,3-Cyclohexanediol, Cis + Trans
EINECSNumber 207-979-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only