missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanediol, 98%, mixture of cis and trans

Catalog No. AC111140250
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111140250 25 g Glass bottle
AC111140100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111140250 Supplier Thermo Scientific Chemicals Supplier No. 111140250
Only null left

CAS: 504-01-8 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 504-01-8
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00039458
InChI Key RLMGYIOTPQVQJR-UHFFFAOYSA-N
Synonym 1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7
PubChem CID 10433
IUPAC Name cyclohexane-1,3-diol
SMILES C1CC(CC(C1)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 30°C
Color Yellow
Boiling Point 246°C to 247°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.497
Linear Formula C6H10(OH)2
Quantity 25 g
Beilstein 06, 740
Formula Weight 116.16
Percent Purity 98%
Physical Form Viscous Liquid After Melting
Chemical Name or Material 1, 3-Cyclohexanediol, 98%
Show More Show Less
EINECSNumber 207-979-5
TSCA TSCA

RUO – Research Use Only