missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Cyclohexanedicarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
$113.59 - $398.51
Chemical Identifiers
| CAS | 3971-31-1 |
|---|---|
| Molecular Formula | C8H12O4 |
| Molecular Weight (g/mol) | 172.18 |
| MDL Number | MFCD00134411 |
| InChI Key | XBZSBBLNHFMTEB-UHFFFAOYSA-N |
| PubChem CID | 107205 |
| IUPAC Name | cyclohexane-1,3-dicarboxylic acid |
| SMILES | C1CC(CC(C1)C(=O)O)C(=O)O |
Chemical Identifiers
| 3971-31-1 | |
| 172.18 | |
| XBZSBBLNHFMTEB-UHFFFAOYSA-N | |
| cyclohexane-1,3-dicarboxylic acid |
| C8H12O4 | |
| MFCD00134411 | |
| 107205 | |
| C1CC(CC(C1)C(=O)O)C(=O)O |
Specifications
| 3971-31-1 | |
| White | |
| MFCD00134411 | |
| XBZSBBLNHFMTEB-UHFFFAOYSA-N | |
| cyclohexane-1,3-dicarboxylic acid | |
| 107205 | |
| ≥98.0% (GC,T) | |
| 1,3-Cyclohexanedicarboxylic Acid (cis- and trans- mixture) |
| 149°C | |
| C8H12O4 | |
| 5 g | |
| C1CC(CC(C1)C(=O)O)C(=O)O | |
| 172.18 | |
| 172.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2685
TSCA : Yes