Learn More
1,3-Cyclohexadiene, 96%, stabilized
CAS: 592-57-4 | C6H8 | 80.13 g/mol
Supplier: Thermo Scientific Chemicals 111100250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1,3-Cyclohexadiene | |
| 592-57-4 | |
| 100.0 | |
| Colorless to Yellow | |
| 80.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4730 to 1.4760 | |
| 25 mL | |
| 1,3-cyclohexadiene, cyclohexadiene, 1,2-dihydrobenzene, unii-jv5w0eg5bp, jv5w0eg5bp, 3-cyclohexen-1,2-ylene, 1,3-cyclohexadien, 1,3 cyclohexadiene, 2,4-cyclohexadiene, cyclohexadiene-1,3 | |
| MGNZXYYWBUKAII-UHFFFAOYSA-N | |
| cyclohexa-1,3-diene | |
| 0.1% BHT | |
| CHEBI:37610 | |
| 96% |
| 96% | |
| 95.0 | |
| -98.0°C | |
| 0.8600g/mL | |
| −6°C | |
| 95% min. (GC) | |
| C6H8 | |
| MFCD00001532 | |
| 0.86 | |
| Solubility in water: immiscible. | |
| C1CC=CC=C1 | |
| 80.13 | |
| 11605 | |
| 80.13 | |
| Liquid |
Chemical Identifiers
| 592-57-4 | |
| 80.13 | |
| MGNZXYYWBUKAII-UHFFFAOYSA-N | |
| 11605 | |
| cyclohexa-1,3-diene |
| C6H8 | |
| MFCD00001532 | |
| 1,3-cyclohexadiene, cyclohexadiene, 1,2-dihydrobenzene, unii-jv5w0eg5bp, jv5w0eg5bp, 3-cyclohexen-1,2-ylene, 1,3-cyclohexadien, 1,3 cyclohexadiene, 2,4-cyclohexadiene, cyclohexadiene-1,3 | |
| CHEBI:37610 | |
| C1CC=CC=C1 |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
GHS Signal Word: Danger
EINECSNumber : 209-764-1
TSCA : TSCA
RUO – Research Use Only