missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexadiene, 96%, stab. with 0.1% BHT

Catalog No. AAA1939414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1939414 25 g
AAA1939406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1939414 Supplier Thermo Scientific Chemicals Supplier No. A1939414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 592-57-4 | C6H8 | 80.13 g/mol

1,3-Cyclohexadiene is used as a hydrogen donor in transfer hydrogenation. It is used in the conversion to benzene. It is useful in the study of proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclohexadiene is used as a hydrogen donor in transfer hydrogenation. It is used in the conversion to benzene. It is useful in the study of proteomics research.

Solubility
Slightly miscible with methanol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.

Chemical Identifiers

CAS 592-57-4
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00001532
InChI Key MGNZXYYWBUKAII-UHFFFAOYSA-N
Synonym 1,3-cyclohexadiene, cyclohexadiene, 1,2-dihydrobenzene, unii-jv5w0eg5bp, jv5w0eg5bp, 3-cyclohexen-1,2-ylene, 1,3-cyclohexadien, 1,3 cyclohexadiene, 2,4-cyclohexadiene, cyclohexadiene-1,3
PubChem CID 11605
ChEBI CHEBI:37610
IUPAC Name cyclohexa-1,3-diene
SMILES C1CC=CC=C1

Specifications

Melting Point -89°C
Density 0.847
Boiling Point 79°C to 81°C
Flash Point −8°C (−144°F)
Odor Unpleasant
Refractive Index 1.474
Quantity 25 g
UN Number UN3295
Beilstein 506024
Solubility Information Slightly miscible with methanol. Immiscible with water.
Formula Weight 80.13
Percent Purity 96%
Chemical Name or Material 1,3-Cyclohexadiene, stabilized with 0.1% BHT
Show More Show Less
Hazard Category H225-H335
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 209-764-1
RTECSNumber GU4702350
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only