missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cycloheptanedione, 97%

Catalog No. AAH5458803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5458803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5458803 Supplier Thermo Scientific Chemicals Supplier No. H5458803

CAS: 1194-18-9 | C7H10O2 | 126.155 g/mol

1,3-Cyclopentanedione was used in the synthesis of chemical probes for selective labeling of sulfuric acid proteins. is a versatile reagent for synthesis of perhydroazulenes, prostaglandin products (biologically active unsaturated 20-carbon fatty acids including a 5-carbon ring, derived enzymatically ), and Knoevenagel products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclopentanedione was used in the synthesis of chemical probes for selective labeling of sulfuric acid proteins. is a versatile reagent for synthesis of perhydroazulenes, prostaglandin products (biologically active unsaturated 2

Chemical Identifiers

CAS 1194-18-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD01863735
InChI Key DBOVMTXPZWVYAQ-UHFFFAOYSA-N
Synonym 1,3-cycloheptanedione, 1 3-cycloheptanedione 97, acmc-20aijy, 1,3-cycloheptandione, pubchem20261, cycloheptan-1,3-dione, cyclohep tane-1,3-dione, cycloheptane-1,3-quinone, cycloheptane-?1,?3-?dione, ksc173k8n
PubChem CID 4072367
IUPAC Name cycloheptane-1,3-dione
SMILES C1CCC(=O)CC(=O)C1

Specifications

Density 1.1
Boiling Point 254°C
Flash Point >110°C (230°F)
Refractive Index 1.483
Quantity 1 g
Solubility Information Soluble in water (5.5g/100 g),and dimethyl formamide (DMF)
Formula Weight 126.15
Percent Purity 97%
Chemical Name or Material 1,3-Cycloheptanedione
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only